COMH42119F28 Combi-Blocks, Inc.

N-(5-Amino-2-hydroxyphenyl)naphthalene-2-carboxamide hydrochloride

  • MDL number MFCD32065745
  • Cas Number : 2365419-29-8
  • Molecular Weight : 314.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(5-amino-2-hydroxyphenyl)-2-naphthamide hydrochloride
Inchi Code 1S/C17H14N2O2.ClH/c18-14-7-8-16(20)15(10-14)19-17(21)13-6-5-11-3-1-2-4-12(11)9-13;/h1-10,20H,18H2,(H,19,21);1H
Inchi Key WGADAAJANZWPBJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4805
Availability details for
Quantity
  • Availability
  • Add to Cart