COMH42119F29 Combi-Blocks, Inc.

4-tert-Butyl-N,N-diisopropylbenzenecarboximidamide hydrochloride

  • MDL number MFCD32065746
  • Cas Number : 2365419-15-2
  • Molecular Weight : 296.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4807
Availability details for
Quantity
  • Availability
  • Add to Cart