COMH42119F2F Combi-Blocks, Inc.

N-(4-Bromo-5-methyl-1,3-thiazol-2-yl)formamide

  • MDL number MFCD32065761
  • Cas Number : 2365418-84-2
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4899
Availability details for
Quantity
  • Availability
  • Add to Cart