COMH42119F30 Combi-Blocks, Inc.

2-Methoxy-3-(2-methoxyethoxy)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD32065765
  • Cas Number : 2377609-09-9
  • Molecular Weight : 309.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1599
Availability details for
Quantity
  • Availability
  • Add to Cart