COMH42119F31 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-[5-methyl-4-(methylsulfanyl)furan-2-yl]-1,3,2-dioxaborolane

  • MDL number MFCD32065766
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1607

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1607
Availability details for
Quantity
  • Availability
  • Add to Cart