COMH42119F32 Combi-Blocks, Inc.

tert-Butyl N-[(tert-butoxy)carbonyl]-N-[3-isopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

  • MDL number MFCD32065767
  • Cas Number : 2377610-78-9
  • Molecular Weight : 461.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PC-1641
Availability details for
Quantity
  • Availability
  • Add to Cart