COMH42119F34 Combi-Blocks, Inc.

Benzyl N-[2,3-difluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

  • MDL number MFCD32065769
  • Cas Number : 2377607-45-7
  • Molecular Weight : 389.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5856
Availability details for
Quantity
  • Availability
  • Add to Cart