COMH42119F3C Combi-Blocks, Inc.

5-Bromo-2,4-dihydroxybenzaldehyde

  • MDL number MFCD20133738
  • Cas Number : 116096-90-3
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-dihydroxybenzaldehyde
Inchi Code 1S/C7H5BrO3/c8-5-1-4(3-9)6(10)2-7(5)11/h1-3,10-11H
Inchi Key YIIXIVBXVLTMHQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9119
Availability details for
Quantity
  • Availability
  • Add to Cart