COMH42119F3F Combi-Blocks, Inc.

6-Amino-1,4-dimethyl-1,4-dihydro-2,3-quinoxalinedione

  • MDL number MFCD06809633
  • Cas Number : 76097-86-4
  • Molecular Weight : 205.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-amino-1,4-dimethyl-1,4-dihydro-2,3-quinoxalinedione
Inchi Code 1S/C10H11N3O2/c1-12-7-4-3-6(11)5-8(7)13(2)10(15)9(12)14/h3-5H,11H2,1-2H3
Inchi Key YXTXXHXCYQXDSU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9126
Availability details for
Quantity
  • Availability
  • Add to Cart