COMH42119F44 Combi-Blocks, Inc.

Benzyl-peg4-acid

  • MDL number MFCD31561129
  • Cas Number : 127457-64-1
  • Molecular Weight : 312.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9158
Availability details for
Quantity
  • Availability
  • Add to Cart