COMH42119F4B Combi-Blocks, Inc.

(2-Chloro-6-piperidin-1-yl-9h-purin-9-yl)acetic acid

  • MDL number MFCD22375173
  • Cas Number : 1383626-14-9
  • Molecular Weight : 295.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9192
Availability details for
Quantity
  • Availability
  • Add to Cart