COMH42119F4C Combi-Blocks, Inc.

Bis-(2-allyloxy-ethyl)-amine

  • MDL number MFCD00464786
  • Cas Number : 210639-26-2
  • Molecular Weight : 185.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9194
Availability details for
Quantity
  • Availability
  • Add to Cart