COMH42119F4E Combi-Blocks, Inc.

1-(9H-Fluoren-9-yl)-4-methyl-3-oxo-2,7,10,13,16-pentaoxa-4-azanonadecan-19-oic acid

  • MDL number MFCD29090721
  • Cas Number : 2170240-98-7
  • Molecular Weight : 501.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9202

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9202
Availability details for
Quantity
  • Availability
  • Add to Cart