COMH42119F58 Combi-Blocks, Inc.

[2-(Biphenyl-4-yloxy)-5-(trifluoromethyl)phenyl]amine hydrochloride

  • MDL number MFCD12128989
  • Cas Number : 904253-69-6
  • Molecular Weight : 365.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9242
Availability details for
Quantity
  • Availability
  • Add to Cart