COMH42119F6F Combi-Blocks, Inc.

1-Benzyl-3-bromo-5-hydrazino-1h-1,2,4-triazole dihydrochloride

  • MDL number MFCD31619034
  • Cas Number : 2279123-31-6
  • Molecular Weight : 341.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9282
Availability details for
Quantity
  • Availability
  • Add to Cart