COMH42119F78 Combi-Blocks, Inc.

N-[3-(Hydrazinocarbonyl)phenyl]-2-methylbenzamide

  • MDL number MFCD02255615
  • Cas Number : 2270909-33-4
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9319

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9319
Availability details for
Quantity
  • Availability
  • Add to Cart