COMH42119F7A Combi-Blocks, Inc.

5-(Hydroxymethyl)pyrimidine-2-carbaldehyde

  • MDL number MFCD25963244
  • Cas Number : 40749-33-5
  • Molecular Weight : 138.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9324
Availability details for
Quantity
  • Availability
  • Add to Cart