COMH42119F7B Combi-Blocks, Inc.

2-(4-(tert-Butyl)-1-(1h-pyrrol-1-yl)cyclohexyl)acetic acid

  • MDL number MFCD23136078
  • Cas Number : 1797082-24-6
  • Molecular Weight : 263.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9327
Availability details for
Quantity
  • Availability
  • Add to Cart