COMH42119F7E Combi-Blocks, Inc.

Methyl 1-(3-methylbenzyl)-1h-1,2,3-triazole-4-carboxylate

  • MDL number MFCD03866364
  • Cas Number : 141072-09-5
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9335
Availability details for
Quantity
  • Availability
  • Add to Cart