COMH42119F89 Combi-Blocks, Inc.

6-Chloro-2,3,4,9-tetrahydro-1h-carbazol-1-amine

  • MDL number MFCD01241897
  • Cas Number : 352553-60-7
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9374
Availability details for
Quantity
  • Availability
  • Add to Cart