COMH42119F8B Combi-Blocks, Inc.

[5-(Methoxycarbonyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyridin-3-yl]acetic acid

  • MDL number MFCD31619038
  • Cas Number : 2279122-94-8
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9382
Availability details for
Quantity
  • Availability
  • Add to Cart