COMH42119F96 Combi-Blocks, Inc.

6-Fluoro-2,3,4,9-tetrahydro-1h-carbazol-1-amine

  • MDL number MFCD21218733
  • Cas Number : 1429901-83-6
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-2,3,4,9-tetrahydro-1H-carbazol-1-ylamine
Inchi Code 1S/C12H13FN2/c13-7-4-5-11-9(6-7)8-2-1-3-10(14)12(8)15-11/h4-6,10,15H,1-3,14H2
Inchi Key JNAQAPNLBDHNMO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9430
Availability details for
Quantity
  • Availability
  • Add to Cart