COMH42119F98 Combi-Blocks, Inc.

2,4,6,2',4',6'-Hexamethyl-biphenyl-3,3'-diamine

  • MDL number MFCD28248815
  • Cas Number : 37055-28-0
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9438
Availability details for
Quantity
  • Availability
  • Add to Cart