COMH42119FA6 Combi-Blocks, Inc.

6-Methyl-4-oxo-3,4-dihydrothieno[3,2-d][1,2,3]triazine-7-carboxylic acid

  • MDL number MFCD14281563
  • Cas Number : 1170546-94-7
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9480
Availability details for
Quantity
  • Availability
  • Add to Cart