COMH42119FA8 Combi-Blocks, Inc.

5-(2-Nitrophenyl)-1h-1,2,4-triazole

  • MDL number MFCD28365624
  • Cas Number : 6219-54-1
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9499
Availability details for
Quantity
  • Availability
  • Add to Cart