COMH42119FCE Combi-Blocks, Inc.

5-Methoxy-1-methyl-1h-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD31537609
  • Cas Number : 2167370-98-9
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9600

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9600
Availability details for
Quantity
  • Availability
  • Add to Cart