COMH42119FD3 Combi-Blocks, Inc.

1,3-Benzodioxole-2-carboxylic acid methyl ester

  • MDL number MFCD09034160
  • Cas Number : 57984-85-7
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9613
Availability details for
Quantity
  • Availability
  • Add to Cart