COMH42119FDF Combi-Blocks, Inc.

Trisulfo-cy5-alkyne

  • MDL number MFCD29052182
  • Cas Number : 2055138-90-2
  • Molecular Weight : 788.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9681
Availability details for
Quantity
  • Availability
  • Add to Cart