COMH42119FE6 Combi-Blocks, Inc.

H-Phe-gly-nh2 hcl

  • MDL number MFCD30475323
  • Cas Number : 38678-61-4
  • Molecular Weight : 257.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9732
Availability details for
Quantity
  • Availability
  • Add to Cart