COMH42119FE7 Combi-Blocks, Inc.

5-Hydrazino-1-(methoxymethyl)-1h-1,2,4-triazole

  • MDL number MFCD31557137
  • Cas Number : 2200882-53-5
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9741
Availability details for
Quantity
  • Availability
  • Add to Cart