COMH42119FED Combi-Blocks, Inc.

Methyl (2e)-2-(aminomethyl)-3-(4-chlorophenyl)acrylate hydrochloride

  • MDL number MFCD31583408
  • Cas Number : 2270915-04-1
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9760
Availability details for
Quantity
  • Availability
  • Add to Cart