COMH42119FFA Combi-Blocks, Inc.

1-[2-Methoxy-5-(trifluoromethoxy)phenyl]-n-methylmethanamine hydrochloride

  • MDL number MFCD31583445
  • Cas Number : 2270905-57-0
  • Molecular Weight : 271.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9796
Availability details for
Quantity
  • Availability
  • Add to Cart