COMH4211A004 Combi-Blocks, Inc.

Bis(m-peg4)-n-oh

  • MDL number MFCD31561147
  • Cas Number : 2182601-79-0
  • Molecular Weight : 413.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N-di(2,5,8,11-tetraoxatridecan-13-yl)hydroxylamine
Inchi Code 1S/C18H39NO9/c1-21-7-9-25-15-17-27-13-11-23-5-3-19(20)4-6-24-12-14-28-18-16-26-10-8-22-2/h20H,3-18H2,1-2H3
Inchi Key SHFFXGVDVNYUOA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9852

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9852
Availability details for
Quantity
  • Availability
  • Add to Cart