COMH4211A010 Combi-Blocks, Inc.

2-[(2,5-Dioxo-1-phenylpyrrolidin-3-yl)thio]benzoic acid, n,n-diethylethanamine (1:1)

  • MDL number MFCD28505118
  • Cas Number : 1993278-28-6
  • Molecular Weight : 428.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9898
Availability details for
Quantity
  • Availability
  • Add to Cart