COMH4211A015 Combi-Blocks, Inc.

3-((2S,3S)-3-Amino-1-(4-chlorophenyl)butan-2-yl)benzonitrile

  • MDL number MFCD11042325
  • Cas Number : 1002752-56-8
  • Molecular Weight : 284.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9931
Availability details for
Quantity
  • Availability
  • Add to Cart