COMH4211A019 Combi-Blocks, Inc.

2-([(4-Fluoro-phenylcarbamoyl)-methyl]-amino)-benzoic acid

  • MDL number MFCD22385577
  • Cas Number : 1394021-66-9
  • Molecular Weight : 288.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-{[2-(4-fluoroanilino)-2-oxoethyl]amino}benzoic acid
Inchi Code 1S/C15H13FN2O3/c16-10-5-7-11(8-6-10)18-14(19)9-17-13-4-2-1-3-12(13)15(20)21/h1-8,17H,9H2,(H,18,19)(H,20,21)
Inchi Key INGFZNGGNPXHRH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8908
Availability details for
Quantity
  • Availability
  • Add to Cart