COMH4211A028 Combi-Blocks, Inc.

5-(Hydroxymethyl)-1-isopropyl-2-piperidinone

  • MDL number MFCD22396869
  • Cas Number : 1824414-80-3
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(hydroxymethyl)-1-isopropyl-2-piperidinone
Inchi Code 1S/C9H17NO2/c1-7(2)10-5-8(6-11)3-4-9(10)12/h7-8,11H,3-6H2,1-2H3
Inchi Key LQPGSKQUROLYLQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Availability details for
Quantity
  • Availability
  • Add to Cart