COMH4211A03B Combi-Blocks, Inc.

[2-(4-bromophenyl)ethyl]diethylamine

  • MDL number MFCD15526812
  • Cas Number : 174769-98-3
  • Molecular Weight : 256.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2804
Availability details for
Quantity
  • Availability
  • Add to Cart