COMH4211A03E Combi-Blocks, Inc.

1-(3-bromophenyl)cyclohexane-1-carbonitrile

  • MDL number MFCD11036802
  • Cas Number : 676138-32-2
  • Molecular Weight : 264.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-2970
Availability details for
Quantity
  • Availability
  • Add to Cart