COMH4211A041 Combi-Blocks, Inc.

2-bromo-N-isopropylaniline

  • MDL number MFCD11144847
  • Cas Number : 156643-24-2
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3106
Availability details for
Quantity
  • Availability
  • Add to Cart