COMH4211A04B Combi-Blocks, Inc.

3-(2-Oxocyclopentyl)propanenitrile

  • MDL number MFCD00015762
  • Cas Number : 4594-77-8
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-3522
Availability details for
Quantity
  • Availability
  • Add to Cart