COMH4211A055 Combi-Blocks, Inc.

[6-(Oxetan-3-yloxy)pyridin-3-yl]boronic acid

  • MDL number MFCD26521353
  • Cas Number : 2377610-41-6
  • Molecular Weight : 195.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4514
Availability details for
Quantity
  • Availability
  • Add to Cart