COMH4211A058 Combi-Blocks, Inc.

5-bromo-N-ethyl-4-fluoro-2-nitroaniline

  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4545
Availability details for
Quantity
  • Availability
  • Add to Cart