COMH4211A059 Combi-Blocks, Inc.

5-Bromo-N-cyclopropyl-4-fluoro-2-nitroaniline

  • MDL number MFCD32065686
  • Cas Number : 1602988-41-9
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HC-4546
Availability details for
Quantity
  • Availability
  • Add to Cart