COMH4211A071 Combi-Blocks, Inc.

2H-Pyrazole-3-carbonitrile

  • MDL number MFCD00107698
  • Cas Number : 36650-74-5
  • Molecular Weight : 93.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2407
Availability details for
Quantity
  • Availability
  • Add to Cart