COMH4211A073 Combi-Blocks, Inc.

8-Bromoimidazo[1,2-c]pyrimidine

  • MDL number MFCD20527742
  • Cas Number : 1049001-84-4
  • Molecular Weight : 198.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2410
Availability details for
Quantity
  • Availability
  • Add to Cart