COMH4211A074 Combi-Blocks, Inc.

3-(4-Nitrophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD27949456
  • Cas Number : 32856-95-4
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2411

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HI-2411
Availability details for
Quantity
  • Availability
  • Add to Cart