COMH4211A0BC Combi-Blocks, Inc.

Benzyl N-[(cyclohexylcarbamoyl)methyl]carbamate

  • MDL number MFCD14869199
  • Cas Number : 3496-41-1
  • Molecular Weight : 290.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-0779
Availability details for
Quantity
  • Availability
  • Add to Cart