COMH4211A0BE Combi-Blocks, Inc.

2-(4-bromophenyl)pyrimidine

  • MDL number MFCD17171418
  • Cas Number : 461702-59-0
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-1066
Availability details for
Quantity
  • Availability
  • Add to Cart