COMH4211A0C3 Combi-Blocks, Inc.

Benzyl 2-[(morpholin-4-yl)carbonyl]pyrrolidine-1-carboxylate

  • Cas Number : 1869468-05-2
  • Molecular Weight : 318.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2169

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2169
Availability details for
Quantity
  • Availability
  • Add to Cart